siRNA Details
Gene Symbol: Hspa1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_212546.4 | NP_997711.1 | 2 | 915 | ||
GenBank mRNA | BC108294.1 | - | 1 | 777 | 7111250 | |
BC108297.1 | - | 2 | 915 | 7115041 |
Gene Symbol: Hspa1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031971.2 | NP_114177.2 | 1995 | |||
GenBank mRNA | L16764.1 | - | 925 |
Gene Symbol: Hspa1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_212504.1 | NP_997669.1 | 1 | 2898 |

More Information
Other Species Targeted

Gene Ontology Categories:
