siRNA Details
Gene Symbol: ABCC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105515.2 | NP_001098985.1 | 2, 3 | 324 | ||
NM_001301829.1 | NP_001288758.1 | 2, 3 | 324 | |||
NM_001301830.1 | NP_001288759.1 | 2, 3 | 324 | |||
NM_005845.4 | NP_005836.2 | 2, 3 | 324 | |||
XM_005254025.2 | XP_005254082.1 | 3, 4 | 384 | |||
XM_017020319.1 | XP_016875808.1 | 3, 4 | 470 | |||
XM_017020320.1 | XP_016875809.1 | 2, 3 | 428 | |||
XM_017020322.1 | XP_016875811.1 | 3, 4 | 333 | |||
GenBank mRNA | AF071202.1 | - | 2, 3 | 307 | ||
AF541977.1 | - | 2, 3 | 197 | |||
AK290317.1 | - | 2, 3 | 324 | |||
AK296247.1 | - | 2, 3 | 311 | |||
AK298953.1 | - | 2, 3 | 288 | |||
AY081219.1 | - | 2, 3 | 213 | |||
AY133678.1 | - | 3, 4 | 366 | |||
AY133679.1 | - | 4, 5 | 479 | |||
AY133680.1 | - | 3, 4 | 326 | |||
AY207008.1 | - | 2, 3 | 197 | |||
BC041560.1 | - | 2, 3 | 294 | 5812052 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
