siRNA Details
Gene Symbol: PFKFB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145443.2 | NP_001138915.1 | 3 | 347 | ||
NM_001282630.2 | NP_001269559.1 | 3 | 460 | |||
NM_001314063.1 | NP_001300992.1 | 3 | 671 | |||
NM_001323016.1 | NP_001309945.1 | 3 | 347 | |||
NM_001323017.1 | NP_001309946.1 | 3 | 671 | |||
NM_004566.3 | NP_004557.1 | 3 | 671 | |||
NR_136554.1 | 3 | 347 | ||||
XM_005252464.1 | XP_005252521.1 | 3 | 671 | |||
XM_011519493.1 | XP_011517795.1 | 3 | 671 | |||
XM_017016325.1 | XP_016871814.1 | 3 | 671 | |||
XM_017016326.1 | XP_016871815.1 | 3 | 671 | |||
XM_017016327.1 | XP_016871816.1 | 3 | 423 | |||
XM_017016328.1 | XP_016871817.1 | 3 | 671 | |||
XM_017016329.1 | XP_016871818.1 | 3 | 671 | |||
GenBank mRNA | AF056320.1 | - | 4 | 331 | ||
AF109735.1 | - | 3 | 401 | |||
AK131307.1 | - | 3 | 307 | |||
AK291263.1 | - | 3 | 616 | |||
AK303039.1 | - | 3 | 280 | |||
AK304450.1 | - | 3 | 392 | |||
BC037258.1 | - | 3 | 272 | |||
BC040482.1 | - | 3 | 616 | 5271907 | ||
D49817.1 | - | 3 | 305 | |||
DC319465.1 | - | 3 | 348 | |||
DC323583.1 | - | 3 | 345 | |||
L77662.1 | - | 3 | 625 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
