siRNA Details
Gene Symbol: Fmr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052804.1 | NP_434691.1 | 4, 5 | 443 | ||
XM_006229530.2 | XP_006229592.1 | 4, 5 | 493 | |||
XM_006229531.2 | XP_006229593.1 | 4, 5 | 493 | |||
XM_006229532.2 | XP_006229594.1 | 4, 5 | 493 | |||
XM_006229535.2 | XP_006229597.1 | 3, 4 | 212 | |||
XM_008773597.1 | XP_008771819.1 | 4, 5 | 325 | |||
GenBank mRNA | AF435434.1 | - | 4, 5 | 262 | ||
AY240947.1 | - | 4, 5 | 443 | |||
U60145.1 | - | 2, 3 | 152 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_008031, AK053701, AK080948, AK053211, AK140343, and others.
Gene Ontology Categories:
