siRNA Details
Gene Symbol: Cast | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053295.2 | NP_445747.2 | 6 | 480 | ||
XM_006231698.2 | XP_006231760.1 | 6 | 888 | |||
XM_006231699.2 | XP_006231761.1 | 6 | 682 | |||
GenBank mRNA | AF346598.1 | - | 4 | 161 | ||
BC070882.1 | - | 6 | 379 | |||
DQ186624.1 | - | 6 | 674 | |||
DQ186626.1 | - | 6 | 674 | |||
Y13588.1 | - | 4 | 173 | |||
Y13590.1 | - | 4 | 173 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
