siRNA Details
Gene Symbol: Ackr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053352.1 | NP_445804.1 | 2 | 1080 | ||
XM_006245479.2 | XP_006245541.1 | 3 | 1274 | |||
XM_008767299.1 | XP_008765521.1 | 2 | 4399 | |||
XM_008767300.1 | XP_008765522.1 | 3 | 3360 | |||
GenBank mRNA | AF118816.1 | - | 2 | 1080 | ||
AJ010828.1 | - | 2 | 1075 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001271607, NM_007722, AK031100, AF000236, BC015254.
Set Membership: |
![]() |
Gene Ontology Categories:
