siRNA Details
Gene Symbol: Atox1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF127137.1 | - | 3 | 159 | ||
AF177671.1 | - | 3 | 238 | |||
BC058458.1 | - | 3 | 247 | 6919265 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
