siRNA Details
Gene Symbol: OPN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014322.2 | NP_055137.2 | 2 | 686 | ||
GenBank mRNA | AF140242.1 | - | 2 | 596 | ||
AF303588.1 | - | 2 | 643 | |||
BC036773.1 | - | 2 | 638 | 5723543 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
