siRNA Details
Gene Symbol: Pgk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053291.3 | NP_445743.2 | 4 | 400 | ||
GenBank mRNA | BC063161.1 | - | 4 | 481 | 6921844 | |
BC087651.1 | - | 4 | 400 | 7310274 | ||
CF976116.1 | - | 4 | 499 | |||
M31788.1 | - | 4 | 436 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
