siRNA Details
Gene Symbol: Pkm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053297.2 | NP_445749.1 | 9 | 1344 | ||
XM_006243187.2 | XP_006243249.1 | 9 | 1574 | |||
XM_006243190.2 | XP_006243252.1 | 10 | 1646 | |||
GenBank mRNA | FQ223721.1 | - | 9 | 1352 | ||
FQ224316.1 | - | 6 | 1002 | |||
X15800.1 | - | 9 | 1346 |
