siRNA Details
Gene Symbol: Kcnj16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053314.2 | NP_445766.2 | 1 | 1286 | ||
XM_006247568.2 | XP_006247630.1 | 2 | 1439 | |||
XM_006247569.2 | XP_006247631.1 | 2 | 1526 | |||
XM_006247570.2 | XP_006247632.1 | 2 | 2104 | |||
XM_008768347.1 | XP_008766569.1 | 2 | 1530 | |||
GenBank mRNA | BC078776.1 | - | 1 | 1286 | 7107402 |
