siRNA Details
Gene Symbol: Degs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053323.2 | NP_445775.2 | 3 | 1007 | ||
XM_006250403.2 | XP_006250465.1 | 3 | 1302 | |||
XM_006250404.2 | XP_006250466.1 | 3 | 1248 | |||
GenBank mRNA | AY036902.1 | - | 3 | 989 | ||
BC083727.1 | - | 3 | 1007 | 7191263 | ||
FQ225269.1 | - | 3 | 1012 |
