siRNA Details
Gene Symbol: Ufd1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053418.2 | NP_445870.1 | 2 | 213 | ||
GenBank mRNA | AF234601.1 | - | 2 | 188 | ||
BC161818.1 | - | 2 | 224 | 9101868 | ||
CK599711.1 | - | 213 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_011672, AK147138, AK010461, BC006630, AK088823, and others.
Gene Ontology Categories:
