siRNA Details
Gene Symbol: Fads1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053445.2 | NP_445897.2 | 1 | 216 | ||
XM_006231075.2 | XP_006231137.1 | 1 | 291 | |||
GenBank mRNA | AB052085.1 | - | 1 | 216 | ||
AF320509.1 | - | 1 | 245 | |||
FQ218734.1 | - | 1 | 260 | |||
HQ909026.1 | - | 1 | 140 |

More Information
Other Species Targeted

Gene Ontology Categories:
