siRNA Details
Gene Symbol: Slc25a14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053501.2 | NP_445953.1 | 6 | 656 | ||
XM_006257531.2 | XP_006257593.1 | 5 | 649 | |||
XM_006257532.1 | XP_006257594.1 | 6 | 693 | |||
XM_006257534.2 | XP_006257596.1 | 6 | 1589 | |||
XM_006257535.1 | XP_006257597.1 | 6 | 511 | |||
XM_008773530.1 | XP_008771752.1 | 5 | 654 | |||
XM_008773531.1 | XP_008771753.1 | 7 | 546 | |||
GenBank mRNA | AB028879.1 | - | 5 | 453 | ||
AF300424.1 | - | 6 | 657 | |||
AJ300165.1 | - | 5 | 462 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
