siRNA Details
Gene Symbol: Wnt4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053402.2 | NP_445854.1 | 4, 5 | 601 | ||
GenBank mRNA | AF188608.1 | - | 4, 5 | 602 | ||
BC098752.1 | - | 4, 5 | 622 | 7369699 |

More Information
Other Species Targeted

Gene Ontology Categories:
