siRNA Details
Gene Symbol: Bnip3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053420.3 | NP_445872.1 | 5 | 597 | ||
GenBank mRNA | AF243515.1 | - | 5 | 561 | ||
AY095482.1 | - | 5 | 561 | |||
BC070958.1 | - | 5 | 555 | 7105707 | ||
BC081690.1 | - | 5 | 535 | 7190121 | ||
CO403082.1 | - | 5 | 633 | |||
FQ213448.1 | - | 4 | 358 | |||
FQ215108.1 | - | 5 | 567 | |||
FQ215127.1 | - | 5 | 567 | |||
FQ215203.1 | - | 5 | 588 | |||
FQ216745.1 | - | 5 | 588 | |||
FQ216768.1 | - | 2 | 174 | |||
FQ220853.1 | - | 5 | 566 | |||
KC660101.1 | - | 4 | 394 |

More Information
Other Species Targeted

Gene Ontology Categories:
