siRNA Details
Gene Symbol: Slc7a8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053442.1 | NP_445894.1 | 2 | 944 | ||
GenBank mRNA | AB024400.1 | - | 2 | 944 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_016972, BC059004, AK044620, Y19022, AF171668.
Gene Ontology Categories:
