siRNA Details
Gene Symbol: MAP4K2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001307990.1 | NP_001294919.1 | 32 | 2673 | ||
NM_004579.4 | NP_004570.2 | 32 | 2697 | |||
XM_011545203.2 | XP_011543505.1 | 31 | 2622 | |||
XM_011545204.2 | XP_011543506.1 | 31 | 2559 | |||
XM_017018093.1 | XP_016873582.1 | 31 | 2535 | |||
XM_017018094.1 | XP_016873583.1 | 31 | 2591 | |||
XM_017018095.1 | XP_016873584.1 | 30 | 2522 | |||
GenBank mRNA | BC039839.1 | - | 32 | 2580 | ||
BC047865.1 | - | 32 | 2660 | 5530523 | ||
U07349.1 | - | 32 | 2626 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
