siRNA Details
Gene Symbol: Fgl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053455.2 | NP_445907.2 | 2 | 1282 | ||
GenBank mRNA | AF323608.2 | - | 2 | 1309 | ||
BC072502.1 | - | 2 | 1231 | |||
BC085357.1 | - | 2 | 1239 | |||
BC107665.1 | - | 2 | 1255 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
