siRNA Details
Gene Symbol: Ppp1r9a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053473.2 | NP_445925.1 | 15 | 3412 | ||
XM_006236057.2 | XP_006236119.1 | 21 | 4198 | |||
XM_006236058.2 | XP_006236120.1 | 21 | 4238 | |||
XM_006236061.2 | XP_006236123.1 | 20 | 4174 | |||
XM_006236063.2 | XP_006236125.1 | 20 | 4132 | |||
XM_006236064.2 | XP_006236126.1 | 19 | 4108 | |||
XM_006236066.2 | XP_006236128.1 | 17 | 3586 | |||
XM_006236068.2 | XP_006236130.1 | 15 | 3495 | |||
GenBank mRNA | U72994.1 | - | 15 | 3416 |

More Information
Other Species Targeted

Gene Ontology Categories:
