siRNA Details
Gene Symbol: Dut | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040271.1 | NP_001035361.1 | 4, 5 | 390 | ||
NM_053592.2 | NP_446044.1 | 5, 6 | 510 | |||
GenBank mRNA | CB325379.1 | - | 4, 5 | 404 | ||
FQ220451.1 | - | 545 | ||||
U64030.2 | - | 5, 6 | 510 |
