siRNA Details
Gene Symbol: Ptprr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113390.1 | NP_001106861.1 | 5 | 759 | ||
NM_053594.2 | NP_446046.2 | 1468 | ||||
GenBank mRNA | BC089186.1 | - | 1 | 250 | 7315399 | |
D38292.1 | - | 1471 | ||||
D64050.1 | - | 5 | 787 | |||
U14914.1 | - | 815 |
