siRNA Details
Gene Symbol: PPARA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001928.2 | NP_001001928.1 | 7 | 1233 | ||
NM_005036.4 | NP_005027.2 | 8 | 1317 | |||
XM_005261655.3 | XP_005261712.1 | 8 | 1610 | |||
XM_005261656.3 | XP_005261713.1 | 9 | 1774 | |||
XM_006724269.3 | XP_006724332.1 | 8 | 1376 | |||
XM_006724270.3 | XP_006724333.1 | 7 | 1282 | |||
XM_011530239.2 | XP_011528541.1 | 9 | 1466 | |||
XM_011530240.2 | XP_011528542.1 | 9 | 1469 | |||
XM_011530241.2 | XP_011528543.1 | 8 | 1374 | |||
XM_011530242.2 | XP_011528544.1 | 7 | 1196 | |||
XM_011530243.2 | XP_011528545.1 | 6 | 1437 | |||
XM_011530244.2 | XP_011528546.1 | 9 | 1473 | |||
XM_011530245.2 | XP_011528547.1 | 7 | 1289 | |||
XM_017028839.1 | XP_016884328.1 | 10 | 1648 | |||
XM_017028840.1 | XP_016884329.1 | 8 | 1266 | |||
GenBank mRNA | AB307690.1 | - | 5 | 1051 | ||
AK289821.1 | - | 8 | 1610 | |||
CR456547.1 | - | 6 | 1187 | |||
CR457435.1 | - | 5 | 1051 | |||
EU395809.1 | - | 4 | 848 | |||
EU650667.1 | - | 5 | 1051 | |||
HQ692862.1 | - | 5 | 1051 | |||
L02932.1 | - | 7 | 1267 | |||
S74349.1 | - | 6 | 1174 | |||
Y07619.1 | - | 5 | 1051 | |||
Y16186.1 | - | 2 | 381 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
