siRNA Details
Gene Symbol: Tpra1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053534.2 | NP_445986.2 | 8 | 751 | ||
XM_006236866.2 | XP_006236928.1 | 7 | 1990 | |||
XM_006236868.2 | XP_006236930.1 | 7 | 791 | |||
GenBank mRNA | AF292116.1 | - | 6 | 557 | ||
FQ228747.1 | - | 8 | 826 |

More Information
Other Species Targeted

