siRNA Details
Gene Symbol: MAP4K2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001307990.1 | NP_001294919.1 | 3 | 293 | ||
NM_004579.4 | NP_004570.2 | 3 | 293 | |||
XM_011545203.2 | XP_011543505.1 | 3 | 287 | |||
XM_011545204.2 | XP_011543506.1 | 3 | 287 | |||
XM_017018093.1 | XP_016873582.1 | 3 | 287 | |||
XM_017018094.1 | XP_016873583.1 | 3 | 230 | |||
XM_017018095.1 | XP_016873584.1 | 3 | 230 | |||
XM_017018096.1 | XP_016873585.1 | 3 | 287 | |||
GenBank mRNA | AK298383.1 | - | 3 | 230 | ||
AK304495.1 | - | 3 | 230 | |||
BC039839.1 | - | 3 | 200 | |||
BC047865.1 | - | 3 | 280 | 5530523 | ||
BM458161.1 | - | 3 | 280 | |||
HY168726.1 | - | 3 | 294 | |||
U07349.1 | - | 3 | 224 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
