siRNA Details
Gene Symbol: ABCC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320032.1 | NP_001306961.1 | 24 | 3794 | ||
NM_005688.3 | NP_005679.2 | 24 | 3679 | |||
XM_005247058.4 | XP_005247115.1 | 24 | 5247 | |||
XM_005247059.4 | XP_005247116.1 | 24 | 3669 | |||
XM_011512314.1 | XP_011510616.1 | 24 | 3570 | |||
XM_011512315.1 | XP_011510617.1 | 24 | 3729 | |||
XM_017005492.1 | XP_016860981.1 | 21 | 4918 | |||
GenBank mRNA | AB005659.1 | - | 18 | 2756 | ||
AB019002.1 | - | 23 | 3545 | |||
AB209103.1 | - | 21 | 3188 | |||
AB209454.1 | - | 24 | 3606 | |||
AF104942.1 | - | 24 | 3618 | |||
AF146074.1 | - | 24 | 3689 | |||
AY196484.1 | - | 22 | 3376 | |||
BC140771.1 | - | 23 | 3555 | 9021639 | ||
BX647245.1 | - | 6 | 804 | |||
U83661.2 | - | 24 | 3612 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
