siRNA Details
Gene Symbol: ABCC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105515.2 | NP_001098985.1 | 15 | 2039 | ||
NM_001301829.1 | NP_001288758.1 | 15 | 2039 | |||
NM_001301830.1 | NP_001288759.1 | 14 | 1814 | |||
NM_005845.4 | NP_005836.2 | 15 | 2039 | |||
XM_005254025.2 | XP_005254082.1 | 16 | 2099 | |||
XM_017020319.1 | XP_016875808.1 | 16 | 2185 | |||
XM_017020320.1 | XP_016875809.1 | 15 | 2143 | |||
XM_017020321.1 | XP_016875810.1 | 5 | 1554 | |||
XM_017020322.1 | XP_016875811.1 | 16 | 2048 | |||
GenBank mRNA | AB208973.1 | - | 13 | 1658 | ||
AF071202.1 | - | 15 | 2022 | |||
AF071203.1 | - | 7 | 677 | |||
AF541977.1 | - | 15 | 1912 | |||
AK290317.1 | - | 15 | 2039 | |||
AK296247.1 | - | 14 | 1801 | |||
AY081219.1 | - | 15 | 1928 | |||
AY133678.1 | - | 16 | 2081 | |||
AY133679.1 | - | 17 | 2194 | |||
AY133680.1 | - | 16 | 2041 | |||
AY207008.1 | - | 15 | 1912 | |||
BC041560.1 | - | 15 | 2009 | 5812052 | ||
BX647118.1 | - | 5 | 414 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
