siRNA Details
Gene Symbol: Dnm1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006248716.2 | XP_006248778.1 | 2, 3 | 401 | ||
XM_006248723.2 | XP_006248785.1 | 2, 3 | 400 | |||
XM_008768841.1 | XP_008767063.1 | 2, 3 | 401 | |||
XM_008768843.1 | XP_008767065.1 | 2, 3 | 401 | |||
GenBank mRNA | AF020209.1 | - | 1, 2 | 99 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
