siRNA Details
Gene Symbol: Dnm1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006248715.2 | XP_006248777.1 | 3, 4 | 470 | ||
XM_006248716.2 | XP_006248778.1 | 3, 4 | 452 | |||
XM_006248717.2 | XP_006248779.1 | 3, 4 | 469 | |||
XM_006248720.2 | XP_006248782.1 | 3, 4 | 469 | |||
XM_006248721.2 | XP_006248783.1 | 3, 4 | 469 | |||
XM_006248723.2 | XP_006248785.1 | 3, 4 | 451 | |||
XM_008768840.1 | XP_008767062.1 | 3, 4 | 470 | |||
XM_008768841.1 | XP_008767063.1 | 3, 4 | 452 | |||
XM_008768843.1 | XP_008767065.1 | 3, 4 | 452 | |||
XM_008768845.1 | XP_008767067.1 | 3, 4 | 469 | |||
XM_008768847.1 | XP_008767069.1 | 3, 4 | 469 | |||
GenBank mRNA | AF020209.1 | - | 2, 3 | 150 | ||
AF020210.1 | - | 2, 3 | 168 | |||
AF132727.1 | - | 3, 4 | 342 |

More Information
Other Species Targeted

Gene Ontology Categories:
