siRNA Details
Gene Symbol: Trpm7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053705.2 | NP_446157.2 | 33 | 5014 | ||
GenBank mRNA | AF375874.1 | - | 3 | 513 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
