siRNA Details
Gene Symbol: Rgs14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053764.1 | NP_446216.1 | 7, 8 | 895 | ||
GenBank mRNA | BC091132.1 | - | 6, 7 | 819 | 7376916 | |
U92279.1 | - | 7, 8 | 895 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
