siRNA Details
Gene Symbol: Cacnb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053851.1 | NP_446303.1 | 2 | 530 | ||
XM_006254298.2 | XP_006254360.1 | 3 | 498 | |||
XM_006254300.2 | XP_006254362.1 | 2 | 439 | |||
XM_006254301.2 | XP_006254363.1 | 2 | 1396 | |||
XM_006254303.2 | XP_006254365.1 | 2 | 695 | |||
XM_006254304.2 | XP_006254366.1 | 2 | 1461 | |||
GenBank mRNA | AF394941.1 | - | 2 | 421 | ||
AF423193.1 | - | 2 | 353 | |||
AY190119.1 | - | 3 | 236 | |||
AY190120.1 | - | 2 | 176 | |||
M80545.1 | - | 2 | 530 |
