siRNA Details
Gene Symbol: Pla2g6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005560.1 | NP_001005560.1 | 2 | 179 | ||
NM_001270796.1 | NP_001257725.1 | 3 | 258 | |||
XM_006241997.2 | XP_006242059.1 | 2 | 1001 | |||
XM_006241998.2 | XP_006242060.1 | 2 | 955 | |||
XM_006241999.2 | XP_006242061.1 | 2 | 1001 | |||
XM_006242000.2 | XP_006242062.1 | 2 | 680 | |||
XM_006242001.2 | XP_006242063.1 | 3 | 312 | |||
XM_006242002.1 | XP_006242064.1 | 2 | 548 | |||
XM_006242003.1 | XP_006242065.1 | 3 | 620 | |||
XM_006242004.2 | XP_006242066.1 | 2 | 241 | |||
XM_006242005.2 | XP_006242067.1 | 2 | 1001 | |||
XM_006242006.2 | XP_006242068.1 | 2 | 679 | |||
XM_008765756.1 | XP_008763978.1 | 3 | 588 | |||
XM_008765757.1 | XP_008763979.1 | 2 | 1001 | |||
GenBank mRNA | BC081916.1 | - | 2 | 179 | 7112407 | |
U51898.1 | - | 2 | 560 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
