siRNA Details
Gene Symbol: Ebf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053820.2 | NP_446272.2 | 4 | 658 | ||
XM_006246117.2 | XP_006246179.1 | 4 | 2239 | |||
XM_006246118.2 | XP_006246180.1 | 4 | 2239 | |||
XM_006246120.2 | XP_006246182.1 | 5 | 1993 | |||
XM_008767631.1 | XP_008765853.1 | 4 | 593 | |||
XM_008767632.1 | XP_008765854.1 | 4 | 1007 | |||
XM_008767633.1 | XP_008765855.1 | 2 | 128 | |||
GenBank mRNA | L24051.1 | - | 4 | 400 |
