siRNA Details
Gene Symbol: Lgals8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053862.2 | NP_446314.2 | 5 | 531 | ||
XM_006254166.2 | XP_006254228.1 | 5 | 797 | |||
XM_006254167.2 | XP_006254229.1 | 5 | 543 | |||
XM_006254168.1 | XP_006254230.1 | 5 | 570 | |||
GenBank mRNA | BC072488.1 | - | 5 | 531 | 7104910 | |
U09824.1 | - | 4 | 511 |
