siRNA Details
Gene Symbol: Dusp6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053883.2 | NP_446335.1 | 2, 3 | 1263 | ||
GenBank mRNA | AY724528.1 | - | 2, 3 | 1264 | ||
BC087003.1 | - | 2, 3 | 1263 | 7124105 | ||
U42627.1 | - | 2, 3 | 1190 | |||
X94185.1 | - | 2, 3 | 1056 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_026268, BC003869, AK088665, AK159502, AK152773, and others.
Gene Ontology Categories:
