siRNA Details
Gene Symbol: Pla2g6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005560.1 | NP_001005560.1 | 15 | 2153 | ||
NM_001270796.1 | NP_001257725.1 | 15 | 2067 | |||
XM_006241997.2 | XP_006242059.1 | 16 | 3047 | |||
XM_006241998.2 | XP_006242060.1 | 16 | 3001 | |||
XM_006241999.2 | XP_006242061.1 | 15 | 2975 | |||
XM_006242000.2 | XP_006242062.1 | 16 | 2726 | |||
XM_006242001.2 | XP_006242063.1 | 17 | 2358 | |||
XM_006242002.1 | XP_006242064.1 | 16 | 2594 | |||
XM_006242003.1 | XP_006242065.1 | 17 | 2666 | |||
XM_006242004.2 | XP_006242066.1 | 16 | 2287 | |||
XM_006242005.2 | XP_006242067.1 | 14 | 2810 | |||
XM_006242006.2 | XP_006242068.1 | 14 | 2488 | |||
XM_008765756.1 | XP_008763978.1 | 17 | 2634 | |||
XM_008765757.1 | XP_008763979.1 | 15 | 2882 | |||
GenBank mRNA | BC081916.1 | - | 15 | 2153 | 7112407 | |
U51898.1 | - | 14 | 2366 |

More Information
Other Species Targeted

Gene Ontology Categories:
