siRNA Details
Gene Symbol: Gps1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270939.1 | NP_001257868.1 | 3 | 252 | ||
NM_053969.3 | NP_446421.3 | 3 | 426 | |||
XM_006247865.1 | XP_006247927.1 | 3 | 421 | |||
XM_006247866.1 | XP_006247928.1 | 3 | 421 | |||
XM_006247867.2 | XP_006247929.1 | 3 | 281 | |||
XM_008768462.1 | XP_008766684.1 | 3 | 421 | |||
XM_008768463.1 | XP_008766685.1 | 3 | 281 | |||
GenBank mRNA | BC061746.1 | - | 3 | 375 | 5599712 | |
X87885.1 | - | 2 | 207 |
