siRNA Details
Gene Symbol: Psmc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_057123.1 | NP_476464.1 | 3 | 269 | ||
GenBank mRNA | BC063157.1 | - | 4 | 281 | 6919865 | |
D50696.1 | - | 3 | 269 | |||
FQ209916.1 | - | 4 | 274 |
