siRNA Details
Gene Symbol: Slc26a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_057127.1 | NP_476468.1 | 4 | 615 | ||
XM_006254784.2 | XP_006254846.1 | 2 | 2591 | |||
XM_006254785.2 | XP_006254847.1 | 3 | 1261 | |||
XM_006254786.2 | XP_006254848.1 | 3 | 2069 | |||
XM_006254787.2 | XP_006254849.1 | 3 | 609 | |||
XM_006254788.2 | XP_006254850.1 | 2 | 474 | |||
GenBank mRNA | D82883.1 | - | 4 | 615 |
