siRNA Details
Gene Symbol: Pdha2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053994.2 | NP_446446.1 | 1 | 660 | ||
GenBank mRNA | BC078757.1 | - | 1 | 660 | 7113421 | |
Z18878.1 | - | 1 | 713 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
