siRNA Details
Gene Symbol: Enpp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_057104.2 | NP_476445.2 | 20 | 2249 | ||
GenBank mRNA | BC081747.1 | - | 20 | 2209 | 7115236 | |
D28560.1 | - | 22 | 2318 | |||
DQ131564.1 | - | 20 | 2249 | |||
FQ233297.1 | - | 6 | 565 |
