siRNA Details
Gene Symbol: Cflar | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033864.2 | NP_001029036.1 | 3 | 495 | ||
NM_057138.2 | NP_476479.1 | 3 | 495 | |||
XM_006244946.2 | XP_006245008.1 | 3 | 1062 | |||
XM_006244947.2 | XP_006245009.1 | 3 | 1169 | |||
XM_006244948.2 | XP_006245010.1 | 2 | 2683 | |||
XM_006244949.2 | XP_006245011.1 | 2 | 498 | |||
XM_006244950.2 | XP_006245012.1 | 1 | 188 | |||
XM_008767071.1 | XP_008765293.1 | 3 | 588 | |||
GenBank mRNA | AF244366.1 | - | 1 | 285 | ||
BC089781.1 | - | 495 | 7376876 |
