siRNA Details
Gene Symbol: Keap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_057152.2 | NP_476493.2 | 2 | 469 | ||
XM_006242591.2 | XP_006242653.1 | 2 | 503 | |||
XM_006242592.2 | XP_006242654.1 | 1 | 1004 | |||
GenBank mRNA | AF304364.1 | - | 1 | 223 | ||
FQ212383.1 | - | 2 | 469 |
