siRNA Details
Gene Symbol: St8sia2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_057156.1 | NP_476497.1 | 2 | 117 | ||
GenBank mRNA | L13445.1 | - | 2 | 117 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_009181, X83562, BC096546.
Gene Ontology Categories:
