siRNA Details
Gene Symbol: Plscr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_057194.1 | NP_476542.1 | 5 | 580 | ||
XM_006243536.1 | XP_006243598.1 | 6 | 655 | |||
XM_006243537.1 | XP_006243599.1 | 6 | 650 | |||
XM_006243538.2 | XP_006243600.1 | 5 | 602 | |||
GenBank mRNA | AY024347.1 | - | 5 | 580 | ||
BC085715.1 | - | 6 | 579 | 7109899 |

More Information
Other Species Targeted

Gene Ontology Categories:
