siRNA Details
Gene Symbol: Socs2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_058208.1 | NP_478115.1 | 2 | 432 | ||
GenBank mRNA | AF075382.1 | - | 2 | 432 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001168656, NM_001168655, NM_007706, NM_001168657, AK137403, and others.
Gene Ontology Categories:
