siRNA Details
Gene Symbol: Rps23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | FQ221811.1 | - | 2 | 183 | ||
FQ222494.1 | - | 2 | 185 | |||
FQ222496.1 | - | 2 | 182 | |||
FQ222641.1 | - | 2 | 183 | |||
FQ223677.1 | - | 2 | 183 | |||
FQ223812.1 | - | 2 | 183 | |||
FQ228659.1 | - | 2 | 183 | |||
FQ228931.1 | - | 2 | 183 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
