siRNA Details
Gene Symbol: Mmp16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080776.1 | NP_542954.1 | 1, 2 | 571 | ||
XM_006237921.2 | XP_006237983.1 | 1, 2 | 637 | |||
GenBank mRNA | D63886.1 | - | 1, 2 | 571 | ||
D85509.1 | - | 1, 2 | 571 |
